CAS RN 773129-45-6
IUPAC Name:
3-[3-chloro-4-(trifluoromethoxy)phenyl]prop-2-enoic acid
Synonyms:
3-[3-chloro-4-(trifluoromethoxy)phenyl]prop-2-enoic acid
Molecular Weight (g/mol):
266.6
Molecular Formula:
C10H6ClF3O3
InChi Key:
SCXWBICVJJGFSL-UHFFFAOYSA-N
SMILES:
OC(=O)C=CC1=CC(Cl)=C(OC(F)(F)F)C=C1
1
–
1
of
1
results
1
3-Chloro-4-(trifluoromethoxy)cinnamic acid, 97%, Thermo Scientific™
CAS: 773129-45-6 Molecular Formula: C10H6ClF3O3 Molecular Weight (g/mol): 266.6 MDL Number: MFCD06205341 InChI Key: SCXWBICVJJGFSL-UHFFFAOYSA-N Synonym: 3-chloro-4-trifluoromethoxy cinnamic acid,2e-3-3-chloro-4-trifluoromethoxy phenyl prop-2-enoic acid,3-3-chloro-4-trifluoromethoxy phenyl acrylic acid PubChem CID: 57367865 IUPAC Name: 3-[3-chloro-4-(trifluoromethoxy)phenyl]prop-2-enoic acid SMILES: C1=CC(=C(C=C1C=CC(=O)O)Cl)OC(F)(F)F