CAS RN 763120-58-7
CAS RN 763120-58-7
IUPAC Name:
(1H-pyrazol-4-yl)boronic acid
Synonyms:
1H-pyrazol-4-ylboronic acid
Molecular Weight (g/mol):
111.9
Molecular Formula:
C3H5BN2O2
InChi Key:
KEZNMOUMHOZFRA-UHFFFAOYSA-N
SMILES:
OB(O)C1=CNN=C1