missing translation for 'onlineSavingsMsg'
Learn More

Pyrazole-4-boronic acid, 97%, Thermo Scientific™

Product Code. 10124204
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10124204 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10124204 Supplier Thermo Scientific Maybridge Supplier No. MAY00042DA
 Restricted Item
Estimated Shipment: 27-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 763120-58-7
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C3H5BN2O2
MDL Number MFCD07368251
Quantity 1 g
Synonym 1h-pyrazole-4-boronic acid, pyrazole-4-boronic acid, 1h-pyrazol-4-yl boronic acid, 4-pyrazoleboronic acid, 1h-pyrazol-4-yl-4-boronic acid, pyrazol-4-ylboronic acid, boronic acid, 1h-pyrazol-4-yl, 1h-pyrazole-4-boronicacid, 4-pyrazole boronic acid, 1h-pyrazol-4-boronic acid
InChI Key KEZNMOUMHOZFRA-UHFFFAOYSA-N
SMILES B(C1=CNN=C1)(O)O
IUPAC Name 1H-pyrazol-4-ylboronic acid
Molecular Weight (g/mol) 111.895
PubChem CID 17750278
Formula Weight 111.9
Percent Purity 97%
Chemical Name or Material Pyrazole-4-boronic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.