accessibility menu, dialog, popup

UserName

CAS RN 7423-93-0

(S)ONH2OHHOCl

CAS RN 7423-93-0

IUPAC Name: (2S)-2-amino-3-(3-chloro-4-hydroxyphenyl)propanoic acid
Synonyms: 3-chlorotyrosine ClY
Molecular Weight (g/mol): 215.63
Molecular Formula: C9H10ClNO3
InChi Key: ACWBBAGYTKWBCD-ZETCQYMHSA-N
SMILES: N[C@@H](CC1=CC=C(O)C(Cl)=C1)C(O)=O
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)