missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-L-tyrosine, MedChemExpress
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
3-Chloro-L-tyrosine is a specific marker of myeloperoxidase-catalyzed oxidation, and is markedly elevated in low density lipoprotein isolated from human atherosclerotic intima.
Specifications
Specifications
| Chemical Name or Material | 3-Chloro-L-tyrosine |
| CAS | 7423-93-0 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C9H10ClNO3 |
| Quantity | 250 mg |
| Solubility Information | H2O : 5 mg/mL (23.19 mM; ultrasonic and warming and heat to 60°C) ∣DMSO : < 1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble or slightly soluble) |
| SMILES | O=C(O)[C@@H](N)CC1=CC=C(O)C(Cl)=C1 |
| Molecular Weight (g/mol) | 215.63 |
| Formula Weight | 215.63 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction