missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Fmoc-L-isoleucine, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | N-Fmoc-L-isoleucine |
| CAS | 71989-23-6 |
| Molecular Formula | C21H23NO4 |
| Quantity | 1 g |
| Synonym | N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-isoleucine, (2S,3S)-2-(((9-Fluorenylmethoxy)carbonyl)amino)-3-methylpentanoic Acid, Fmoc-L-Ile-OH, N-(9-Fluorenylmethoxycarbonyl)-L-isoleucine, N-(9-Fluorenylmethoxycarbonyl)isoleucine, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-isoleucine, NSC 334289 |
| InChI Formula | InChI=1S/C21H23NO4/c1-3-13(2)19(20(23)24)22-21(25)26-12-18-16-10-6-4-8-14(16)15-9-5-7-11-17(15)18/h4-11,13,18-19H,3,12H2,1-2H3,(H,22,25)(H,23,24)/t13-,19-/m0/s1 |
| SMILES | CC[C@H](C)[C@H](NC(=O)OCC1c2ccccc2c3ccccc13)C(=O)O |
| Molecular Weight (g/mol) | 353.41 |
| Formula Weight | 353.1627 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction