accessibility menu, dialog, popup

UserName

CAS RN 71680-92-7

H3CONHNH2S

CAS RN 71680-92-7

IUPAC Name: (4-acetylphenyl)thiourea
Synonyms: 4-acetylphenylthiourea
Molecular Weight (g/mol): 194.25
Molecular Formula: C9H10N2OS
InChi Key: VVIUKYOXYSWCOF-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(NC(N)=S)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results