accessibility menu, dialog, popup

UserName

CAS RN 71119-11-4

H3CCH3HNNHOHON

CAS RN 71119-11-4

IUPAC Name: 2-(2-hydroxy-3-{[1-(1H-indol-3-yl)-2-methylpropan-2-yl]amino}propoxy)benzonitrile
Synonyms: bucindolol 布新洛尔
Molecular Weight (g/mol): 363.46
Molecular Formula: C22H25N3O2
InChi Key: FBMYKMYQHCBIGU-UHFFFAOYNA-N
SMILES: CC(C)(CC1=CNC2=CC=CC=C12)NCC(O)COC1=CC=CC=C1C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results