missing translation for 'onlineSavingsMsg'
Learn More

Bucindolol, MedChemExpress

Product Code. 16426537
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16426537 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16426537 Supplier Medchem Express Supplier No. HY103214
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Bucindolol is a β1-adrenergic receptor blocker, with intrinsic sympathomimetic activity, used in the research of heart failure.

Bucindolol is a β1-adrenergic receptor blocker, with intrinsic sympathomimetic activity, used in the research of heart failure.

Specifications

Chemical Name or Material Bucindolol
CAS 71119-11-4
Color White
Purity Grade Notes Research
Molecular Formula C22H25N3O2
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (275.14 mM; Need ultrasonic)
SMILES N#CC1=CC=CC=C1OCC(O)CNC(C)(C)CC2=CNC3=C2C=CC=C3
Molecular Weight (g/mol) 363.45
Formula Weight 363.45
Percent Purity 99.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.