CAS RN 69-25-0
CAS RN 69-25-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
MedChemExpress Eledoisin
MedChemExpress Eledoisin (Eledone peptide) is a specific agonist of NK2 and NK3 receptors.
2
MedChemExpress Eledoisin
MedChemExpress Eledoisin (Eledone peptide) is a specific agonist of NK2 and NK3 receptors.
3
MedChemExpress Eledoisin
MedChemExpress Eledoisin (Eledone peptide) is a specific agonist of NK2 and NK3 receptors.