missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Eledoisin

Product Code. 16422609
Encompass_Preferred
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 16422609

Brand: MedChemExpress HYP0006

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Eledoisin (Eledone peptide) is a specific agonist of NK2 and NK3 receptors.

Specifications

Purity 98.73%
CAS 69-25-0
Content And Storage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 1188.4 g/mol
Product Type Peptides
Quantity 1 mg
Shipping Condition Room temperature
Solubility H2O : 18.15 mg/mL (15.27 mM∣Need ultrasonic and warming)
Synonym Eledone peptide
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.