CAS RN 676128-62-4
CAS RN 676128-62-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
RSV604 racemate, MedChemExpress
MedChemExpress RSV604 (A-60444) racemate is a racemic mixture, shows less potency against strains of respiratory syncytial virus (RSV) than the S-isomer.