missing translation for 'onlineSavingsMsg'
Learn More

RSV604 racemate, MedChemExpress

Product Code. 16465468
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16465468

Brand: Medchem Express HY12993A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress RSV604 (A-60444) racemate is a racemic mixture, shows less potency against strains of respiratory syncytial virus (RSV) than the S-isomer.

RSV604 (A-60444) racemate is a racemic mixture, shows less potency against strains of respiratory syncytial virus (RSV) than the S-isomer.

Specifications

Chemical Name or Material RSV604 racemate
CAS 676128-62-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H17FN4O2
Quantity 10 mg
Synonym A-60444 racemate
Solubility Information DMSO : 50 mg/mL (128.74 mM; Need ultrasonic)
SMILES O=C1C(NC(NC2=CC=CC=C2F)=O)N=C(C3=CC=CC=C3)C4=CC=CC=C4N1
Molecular Weight (g/mol) 388.39
Formula Weight 388.39
Percent Purity 98.6%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.