accessibility menu, dialog, popup

UserName

CAS RN 67515-57-5

H2NOFFFF

CAS RN 67515-57-5

IUPAC Name: 4-fluoro-3-(trifluoromethyl)benzamide
Synonyms: 4-fluoro-3-(trifluoromethyl)benzamide
Molecular Weight (g/mol): 207.13
Molecular Formula: C8H5F4NO
InChi Key: UUOVPAFAXSKGMP-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=C(F)C(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results