CAS RN 67483-13-0
CAS RN 67483-13-0
IUPAC Name:
disodium 5-isothiocyanato-2-[(1E)-2-(4-isothiocyanato-2-sulfonatophenyl)ethenyl]benzene-1-sulfonate
Synonyms:
disodium 5-isothiocyanato-2-[(1E)-2-(4-isothiocyanato-2-sulfonatophenyl)ethenyl]benzenesulfonate
Molecular Weight (g/mol):
498.47
Molecular Formula:
C16H8N2Na2O6S4
InChi Key:
GEPAYBXVXXBSKP-SEPHDYHBSA-L
SMILES:
[Na+].[Na+].[O-]S(=O)(=O)C1=CC(=CC=C1\C=C\C1=CC=C(C=C1S([O-])(=O)=O)N=C=S)N=C=S
1
–
2
of
2
results
2
MedChemExpress DIDS sodium salt
MedChemExpress DIDS sodium salt (MDL101114ZA) is a dual ABCA1 and VDAC1 inhibitor.