missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Diisothiocyano-2,2'-stilbenedisulfonic Acid, Disodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | 4,4'-Diisothiocyano-2,2'-stilbenedisulfonic acid disodium salt |
| CAS | 67483-13-0 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C16 H8 N2 O6 S4 . 2 Na |
| Quantity | 100 mg |
| Synonym | DIDS |
| InChI Formula | InChI=1S/C16H10N2O6S4.2Na/c19-27(20,21)15-7-13(17-9-25)5-3-11(15)1-2-12-4-6-14(18-10-26)8-16(12)28(22,23)24;;/h1-8H,(H,19,20,21)(H,22,23,24);;/q;2*+1/p-2/b2-1+;; |
| SMILES | [Na+].[Na+].[O-]S(=O)(=O)c1cc(ccc1\C=C\c2ccc(cc2S(=O)(=O)[O-])N=C=S)N=C=S |
| IUPAC Name | disodium;5-isothiocyanato-2-[(E)-2-(4-isothiocyanato-2-sulfonatophenyl)ethenyl]benzenesulfonate |
| Molecular Weight (g/mol) | 498.48 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?