missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress DIDS sodium salt

Product Code. 16444667
Encompass_Preferred
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 16444667

Brand: MedChemExpress HYD0086

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress DIDS sodium salt (MDL101114ZA) is a dual ABCA1 and VDAC1 inhibitor.

Specifications

Quantity 50 mg
Storage Requirements -20°C, sealed storage, away from moisture ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture)
Solubility DMSO : 50 mg/mL (100.30 mM; Need ultrasonic) ∣ H2O : 33.33 mg/mL (66.86 mM; Need ultrasonic)
Shipping Condition Room temperature
CAS 67483-13-0
Molecular Weight (g/mol) 498.48 g/mol
Synonym MDL101114ZA
Target HBV
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.