CAS RN 67292-34-6
CAS RN 67292-34-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dichloro[1,1'-bis(diphenylphosphino)ferrocene]nickel(II), 99%, Thermo Scientific Chemicals
CAS: 67292-34-6 Molecular Formula: C34H28Cl2FeNiP2 Molecular Weight (g/mol): 683.98 MDL Number: MFCD00270284 Synonym: [1,1'-Bis(diphenylphosphino)ferrocene]nickel(II) chloride; NiCl2(dppf) IUPAC Name: Dichloro[1,1'-bis(diphenylphosphino)ferrocene]nickel(II) SMILES: [Fe].Cl[Ni]Cl.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1.c1ccc(c1)P(C1=CC=CC=C1)C1=CC=CC=C1