CAS RN 671225-23-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Prednisolone 17-(Ethyl Carbonate) 21-Acetate, TRC
CAS: 671225-23-3 Molecular Formula: C26 H34 O8 Molecular Weight (g/mol): 474.543 Synonym: Pregna-1,4-diene-3,20-dione, 21-(acetyloxy)-17-[(ethoxycarbonyl)oxy]-11-hydroxy-, (11β)- (9CI),Prednisolone 17-(ethyl carbonate) 21-acetate,11β-Hydroxy-3,20-dioxopregna-1,4-dien-17,21-diyl 21-Acetate 17-(Ethylcarbonate),Prednisolone 21-Acetate 17-Ethyl IUPAC Name: [2-[(8S,9S,10R,11S,13S,14S,17R)-17-ethoxycarbonyloxy-11-hydroxy-10,13-dimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate SMILES: CCOC(=O)O[C@@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3[C@@H](O)C[C@]12C)C(=O)COC(=O)C