missing translation for 'onlineSavingsMsg'
Learn More

Prednisolone 17-(Ethyl Carbonate) 21-Acetate, TRC

Product Code. 30134361
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134361 500 mg 500mg
17940515 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134361 Supplier Toronto Research Chemicals Supplier No. TRCP703725500MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 671225-23-3
Molecular Formula C26 H34 O8
Molecular Weight (g/mol) 474.543
Synonym Pregna-1,4-diene-3,20-dione, 21-(acetyloxy)-17-[(ethoxycarbonyl)oxy]-11-hydroxy-, (11β)- (9CI), Prednisolone 17-(ethyl carbonate) 21-acetate, 11β-Hydroxy-3,20-dioxopregna-1,4-dien-17,21-diyl 21-Acetate 17-(Ethylcarbonate), Prednisolone 21-Acetate 17-Ethyl
IUPAC Name [2-[(8S,9S,10R,11S,13S,14S,17R)-17-ethoxycarbonyloxy-11-hydroxy-10,13-dimethyl-3-oxo-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
SMILES CCOC(=O)O[C@@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3[C@@H](O)C[C@]12C)C(=O)COC(=O)C

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C26H34O8/c1-5-32-23(31)34-26(21(30)14-33-15(2)27)11-9-19-18-7-6-16-12-17(28)8-10-24(16,3)22(18)20(29)13-25(19,26)4/h8,10,12,18-20,22,29H,5-7,9,11,13-14H2,1-4H3/t18-,19-,20-,22+,24-,25-,26-/m0/s1
Formula Weight 474.225
Chemical Name or Material Prednisolone 21-Acetate 17-Ethylcarbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.