accessibility menu, dialog, popup

UserName

CAS RN 6665-67-4

HOOOHO

CAS RN 6665-67-4

IUPAC Name: 5-hydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one
Synonyms: 5-hydroxy-2-(4-hydroxyphenyl)chromen-4-one
Molecular Weight (g/mol): 254.24
Molecular Formula: C15H10O4
InChi Key: OKRNDQLCMXUCGG-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C1=CC(=O)C2=C(O)C=CC=C2O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results