accessibility menu, dialog, popup

UserName

CAS RN 63010-71-9

FONH

CAS RN 63010-71-9

IUPAC Name: 8-fluoro-1,4-dihydroquinolin-4-one
Synonyms: 8-fluoro-1H-quinolin-4-one
Molecular Weight (g/mol): 163.15
Molecular Formula: C9H6FNO
InChi Key: HTELWJVKZKSAQI-UHFFFAOYSA-N
SMILES: FC1=C2NC=CC(=O)C2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results