accessibility menu, dialog, popup

UserName

CAS RN 6290-17-1

H3COOH3COCH3O

CAS RN 6290-17-1

IUPAC Name: ethyl 2-(2,4-dimethyl-1,3-dioxolan-2-yl)acetate
Synonyms: ethyl 2-(2,4-dimethyl-1,3-dioxolan-2-yl)acetate
Molecular Weight (g/mol): 188.22
Molecular Formula: C9H16O4
InChi Key: GSIXJEIRJVOUFB-UHFFFAOYNA-N
SMILES: CCOC(=O)CC1(C)OCC(C)O1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results