accessibility menu, dialog, popup

UserName

H3CONOOCH3O

CAS RN 6277-38-9

IUPAC Name: 1-(4-methoxy-3-nitrophenyl)ethan-1-one
Synonyms: 1-(4-methoxy-3-nitrophenyl)ethanone
Molecular Weight (g/mol): 195.17
Molecular Formula: C9H9NO4
InChi Key: VXLKYQQBEPCMJE-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1[N+]([O-])=O)C(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results