missing translation for 'onlineSavingsMsg'
Learn More

4'-Methoxy-3'-nitroacetophenone, 98+%, Thermo Scientific™

Product Code. 11396806
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11396806 10 g 10g
11306816 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11396806 Supplier Thermo Scientific Alfa Aesar Supplier No. L04149.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6277-38-9
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.17
MDL Number MFCD00017023
InChI Key VXLKYQQBEPCMJE-UHFFFAOYSA-N
Synonym 1-4-methoxy-3-nitrophenyl ethanone, 1-4-methoxy-3-nitrophenyl ethan-1-one, 4-methoxy-3-nitroacetophenone, 3'-nitro-4'-methoxyacetophenone, 4'-methoxy-3'-nitroacetophenone, 4-acetyl-2-nitroanisole, 4-methoxy-3-nitro-acetophenone, 1-acetyl-4-methoxy-3-nitrobenzene, 1-4-methoxy-3-nitrophenyl-ethanone
PubChem CID 80470
IUPAC Name 1-(4-methoxy-3-nitrophenyl)ethanone
SMILES COC1=CC=C(C=C1[N+]([O-])=O)C(C)=O

Specifications

Melting Point 97°C to 99°C
Quantity 10 g
Beilstein 646069
Formula Weight 195.18
Percent Purity ≥98%
Chemical Name or Material 4'-Methoxy-3'-nitroacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.