accessibility menu, dialog, popup

UserName

ONN

CAS RN 6141-98-6

IUPAC Name: benzo[c]cinnolin-5-ium-5-olate
Synonyms: benzo[c]cinnolin-5-ium-5-olate
Molecular Weight (g/mol): 196.21
Molecular Formula: C12H8N2O
InChi Key: JFIBBBQSFGWTPH-UHFFFAOYSA-N
SMILES: [O-][N+]1=NC2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results