missing translation for 'onlineSavingsMsg'
Learn More

Benzo[c]cinnoline N-oxide, 95%, Thermo Scientific™

Product Code. 11415117
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
Product Code. Quantity unitSize
11415117 5 g 5g
1 options
This item is not returnable. View return policy

Product Code. 11415117

Brand: Thermo Scientific Alfa Aesar B22547.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6141-98-6
Molecular Formula C12H8N2O
Molecular Weight (g/mol) 196.209
MDL Number MFCD00014475
InChI Key JFIBBBQSFGWTPH-UHFFFAOYSA-N
Synonym benzo c cinnoline n-oxide, benzo c cinnoline 5-oxide, benzo c cinnoline-n-oxide, benzocinnoline n-oxide, benzo c cinnoline, 5-oxide, 5-oxidobenzo c cinnolin-5-ium, benzo c cinnolin-5-ium-5-olate
PubChem CID 72826
IUPAC Name 5-oxidobenzo[c]cinnolin-5-ium
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3[N+](=N2)[O-]

Specifications

Melting Point 138°C to 141°C
Odor Odorless
Quantity 5 g
Beilstein 8732
Formula Weight 196.21
Percent Purity 95%
Chemical Name or Material Benzo[c]cinnoline N-oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.