accessibility menu, dialog, popup

UserName

CAS RN 61350-00-3

NOHNCl

CAS RN 61350-00-3

IUPAC Name: N-(3-chlorophenyl)pyridine-2-carboxamide
Synonyms: N-(3-chlorophenyl)pyridine-2-carboxamide
Molecular Weight (g/mol): 232.67
Molecular Formula: C12H9ClN2O
InChi Key: SUYUTNCKIOLMAJ-UHFFFAOYSA-N
SMILES: ClC1=CC=CC(NC(=O)C2=CC=CC=N2)=C1
Grade 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results