missing translation for 'onlineSavingsMsg'
Learn More

VU0364770, MedChemExpress

Product Code. p-200097571
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16484386

Brand: Medchem Express HY100588

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress VU0364770 is a selective and potent positive allosteric modulator (PAM) of mGlu4.

VU0346770 exhibits EC50s of 290 nM and 1.1 μM at rat mGlu4 and human mGlu4 receptor, respectively. VU0364770 exhibits antagonist activity at mGlu5 with a potency of 17.9 μM and PAM activity at mGlu6 with a potency of 6.8 μM. VU0364770 also possesses activity at MAO with Ki values of 8.5 and 0.72 μM for human MAO-A and human MAO-B, respectively.

Specifications

Chemical Name or Material VU0364770
CAS 61350-00-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C12H9ClN2O
Quantity 100 mg
Solubility Information DMSO : ≥ 100 mg/mL (429.79 mM)
SMILES O=C(C1=NC=CC=C1)NC2=CC=CC(Cl)=C2
Molecular Weight (g/mol) 232.67
Formula Weight 232.67
Percent Purity 99.57%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.