accessibility menu, dialog, popup

UserName

CAS RN 604-33-1

CH3OO(S)(R)CH3(S)(R)CH3(R)(S)(S)(R)CH3CH3CH3(Z)(Z)

CAS RN 604-33-1

IUPAC Name: (1R,3aS,3bS,7S,9aR,9bS,11aR)-9a,11a-dimethyl-1-[(2R)-6-methylheptan-2-yl]-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-yl (9Z,12Z)-octadeca-9,12-dienoate
Synonyms: cholesteryl linoleate CLOH cholesteryl linolelaidate 18:2 cholesteryl ester
Molecular Weight (g/mol): 649.1
Molecular Formula: C45H76O2
InChi Key: NAACPBBQTFFYQB-LJAITQKLSA-N
SMILES: CCCCC\C=C/C\C=C/CCCCCCCC(=O)O[C@H]1CC[C@]2(C)[C@H]3CC[C@]4(C)[C@H](CC[C@H]4[C@@H]3CC=C2C1)[C@H](C)CCCC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results