accessibility menu, dialog, popup

UserName

CAS RN 6018-41-3

CH3OOOO

CAS RN 6018-41-3

IUPAC Name: methyl 2-oxo-2H-pyran-5-carboxylate
Synonyms: methyl 6-oxopyran-3-carboxylate
Molecular Weight (g/mol): 154.12
Molecular Formula: C7H6O4
InChi Key: HHWWWZQYHPFCBY-UHFFFAOYSA-N
SMILES: COC(=O)C1=COC(=O)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results