missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Coumalate, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Methyl Coumalate |
| CAS | 6018-41-3 |
| Molecular Formula | C7 H6 O4 |
| Quantity | 1 g |
| Synonym | 2H-Pyran-5-carboxylic acid, 2-oxo-, methyl ester, Glutaconic acid, 4-(hydroxymethylene)-, Δ-lactone, methyl ester (7CI), 5-(Carbomethoxy)-2-pyrone, 5-(Methoxycarbonyl)-2-pyrone, Coumalic acid methyl ester, Cumalic acid methyl ester, Methyl 2-oxo-2H-pyran-5-carboxylate, Methyl 2-pyrone-5-carboxylate, Methyl coumalate, Methyl cumalate, NSC 137387, NSC 34627 |
| InChI Formula | InChI=1S/C7H6O4/c1-10-7(9)5-2-3-6(8)11-4-5/h2-4H,1H3 |
| SMILES | COC(=O)C1=COC(=O)C=C1 |
| IUPAC Name | methyl 6-oxopyran-3-carboxylate |
| Molecular Weight (g/mol) | 154.12 |
| Formula Weight | 154.0266 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction