missing translation for 'onlineSavingsMsg'
Learn More

Methyl Coumalate, TRC

Product Code. 30466385
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 30466385

Brand: Toronto Research Chemicals TRCM2945305G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl Coumalate
CAS 6018-41-3
Molecular Formula C7 H6 O4
Quantity 5 g
Synonym 2H-Pyran-5-carboxylic acid, 2-oxo-, methyl ester, Glutaconic acid, 4-(hydroxymethylene)-, Δ-lactone, methyl ester (7CI), 5-(Carbomethoxy)-2-pyrone, 5-(Methoxycarbonyl)-2-pyrone, Coumalic acid methyl ester, Cumalic acid methyl ester, Methyl 2-oxo-2H-pyran-5-carboxylate, Methyl 2-pyrone-5-carboxylate, Methyl coumalate, Methyl cumalate, NSC 137387, NSC 34627
InChI Formula InChI=1S/C7H6O4/c1-10-7(9)5-2-3-6(8)11-4-5/h2-4H,1H3
SMILES COC(=O)C1=COC(=O)C=C1
IUPAC Name methyl 6-oxopyran-3-carboxylate
Molecular Weight (g/mol) 154.12
Formula Weight 154.0266
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.