CAS RN 60133-18-8
CAS RN 60133-18-8
IUPAC Name:
5-{2-[1-(6-hydroxy-5,6-dimethylhept-3-en-2-yl)-7a-methyl-octahydro-1H-inden-4-ylidene]ethylidene}-4-methylidenecyclohexane-1,3-diol
Synonyms:
5-{2-[1-(6-hydroxy-5,6-dimethylhept-3-en-2-yl)-7a-methyl-hexahydro-1H-inden-4-ylidene]ethylidene}-4-methylidenecyclohexane-1,3-diol
Molecular Weight (g/mol):
428.66
Molecular Formula:
C28H44O3
InChi Key:
ZGLHBRQAEXKACO-UHFFFAOYNA-N
SMILES:
CC(C=CC(C)C(C)(C)O)C1CCC2C(CCCC12C)=CC=C1CC(O)CC(O)C1=C
1
–
4
of
4
results
1
1alpha,25-Dihydroxy Vitamin D2, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
1alpha,25-Dihydroxy Vitamin D2, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
1alpha,25-Dihydroxy Vitamin D2, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Ercalcitriol, MedChemExpress
MedChemExpress Ercalcitriol (1α,25-Dihydroxy Vitamin D2) is an active metabolite of vitamin D2.