missing translation for 'onlineSavingsMsg'
Learn More

Ercalcitriol, MedChemExpress

Product Code. 16480498
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16480498 5 mg 5mg
16468038 10 mg 10mg
16463746 25 mg 25mg
16401929 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16480498 Supplier Medchem Express Supplier No. HY32350
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Ercalcitriol (1α,25-Dihydroxy Vitamin D2) is an active metabolite of vitamin D2.

Ercalcitriol (1α,25-Dihydroxy Vitamin D2) is an active metabolite of vitamin D2.

Specifications

Chemical Name or Material Ercalcitriol
CAS 60133-18-8
Color White
Purity Grade Notes Research
Molecular Formula C28H44O3
Quantity 5 mg
Synonym 1α,25-Dihydroxy Vitamin D2
Solubility Information DMSO : ≥ 50 mg/mL (116.65 mM)
SMILES C=C1[C@H](C[C@@H](C/C1=C/C=C2[C@]3([C@@](C)([C@H](CC3)[C@@H](/C=C/[C@@H](C(C)(O)C)C)C)CCC/2)[H])O)O
Molecular Weight (g/mol) 428.65
Formula Weight 428.65
Percent Purity 95.37%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.