accessibility menu, dialog, popup

UserName

CAS RN 60-81-1

HO(R)O(S)OOHOHOOH(R)OH(S)HO(S)HO

CAS RN 60-81-1

IUPAC Name: 1-(2,4-dihydroxy-6-{[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}phenyl)-3-(4-hydroxyphenyl)propan-1-one
Synonyms: phlorizin phloridzin phloretin 2'-glucoside phlorhizin phlorizoside floridzin phlorrhizin
Molecular Weight (g/mol): 436.41
Molecular Formula: C21H24O10
InChi Key: IOUVKUPGCMBWBT-QNDFHXLGSA-N
SMILES: OC[C@H]1O[C@@H](OC2=CC(O)=CC(O)=C2C(=O)CCC2=CC=C(O)C=C2)[C@H](O)[C@@H](O)[C@@H]1O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results