missing translation for 'onlineSavingsMsg'
Learn More

Phlorizin, MedChemExpress

Product Code. 16482499
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16482499

Brand: Medchem Express HYN0143

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Phlorizin is a non-selective SGLT inhibitor with Kis of 300 and 39 nM for hSGLT1 and hSGLT2, respectively. Phlorizin is also a Na+/K+-ATPase inhibitor.

Phlorizin is a non-selective SGLT inhibitor with Kis of 300 and 39 nM for hSGLT1 and hSGLT2, respectively. Phlorizin is also a Na+/K+-ATPase inhibitor.

Specifications

Chemical Name or Material Phlorizin
CAS 60-81-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H24O10
Quantity 1 g
Synonym Floridzin NSC 2833
Solubility Information DMSO : ≥ 50 mg/mL (114.57 mM) ∣H2O : 1 mg/mL (2.29 mM; Need ultrasonic)
SMILES O=C(C1=C(O)C=C(O)C=C1O[C@H]2[C@@H]([C@H]([C@@H]([C@@H](CO)O2)O)O)O)CCC3=CC=C(O)C=C3
Molecular Weight (g/mol) 436.41
Formula Weight 436.41
Percent Purity 99.82%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.