accessibility menu, dialog, popup

UserName

CH3HNOOCH3OHOOOCH3

CAS RN 599150-20-6

IUPAC Name: 2-methoxyethyl 4-(3-hydroxyphenyl)-7-(2-methoxyphenyl)-2-methyl-5-oxo-1,4,5,6,7,8-hexahydroquinoline-3-carboxylate
Synonyms: 2-methoxyethyl 4-(3-hydroxyphenyl)-7-(2-methoxyphenyl)-2-methyl-5-oxo-4,6,7,8-tetrahydro-1H-quinoline-3-carboxylate
Molecular Weight (g/mol): 463.53
Molecular Formula: C27H29NO6
InChi Key: ZXSWZQSYZYMZKS-UHFFFAOYNA-N
SMILES: COCCOC(=O)C1=C(C)NC2=C(C1C1=CC(O)=CC=C1)C(=O)CC(C2)C1=CC=CC=C1OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results