missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HPI 1, Tocris Bioscience™
Description
Hedgehog (Hh) signaling inhibitor. Inhibits Sonic hedgehog (Shh)-, SAG- and Gli-induced Hh pathway activation in Shh-LIGHT2 cells (IC50 values are 1.5, 1.5, 4 and 6 μM for Shh-, SAG-, Gli2- and Gli1-induced activation). Also inhibits Hh pathway activation in SmoM2-LIGHT cells (IC50 = 2.5 μM); inhibits the proliferation of cerebellar granule neuron precursors expressing SmoM2. Does not inhibit Wnt signaling.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 463.52 |
| Percent Purity | >95% |
| Chemical Name or Material | HPI 1 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?