CAS RN 59798-30-0
CAS RN 59798-30-0
IUPAC Name:
sodium 6-{2-[(3-methanesulfonyl-2-oxoimidazolidine-1-carbonyl)amino]-2-phenylacetamido}-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Synonyms:
sodium 6-[2-(3-methanesulfonyl-2-oxoimidazolidine-1-carbonylamino)-2-phenylacetamido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Molecular Weight (g/mol):
561.56
Molecular Formula:
C21H24N5NaO8S2
InChi Key:
GTGQRSIMEUWHPA-UHFFFAOYNA-M
SMILES:
[Na+].CC1(C)SC2C(NC(=O)C(NC(=O)N3CCN(C3=O)S(C)(=O)=O)C3=CC=CC=C3)C(=O)N2C1C([O-])=O
1
–
1
of
1
results
1
Mezlocillin sodium salt, Thermo Scientific™
CAS: 59798-30-0 Molecular Formula: C21H25N5NaO8S2 Molecular Weight (g/mol): 562.568 MDL Number: MFCD07793332 InChI Key: SGVORSZSYKDVFT-KZHFGBRHSA-N PubChem CID: 123133909 IUPAC Name: (2R,5S,6S)-3,3-dimethyl-6-[[(2R)-2-[(3-methylsulfonyl-2-oxoimidazolidine-1-carbonyl)amino]-2-phenylacetyl]amino]-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;sodium SMILES: CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)NC(=O)N4CCN(C4=O)S(=O)(=O)C)C(=O)O)C.[Na]