Learn More
Mezlocillin sodium salt, Thermo Scientific™
A broad spectrum antibiotic
Brand:  Thermo Scientific Alfa Aesar J66306.MC
Quantity 100mg
 View more versions of this product
Product Code. 15477609
  • £148.00 / 100mg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Identifiers
59798-30-0
562.568
SGVORSZSYKDVFT-KZHFGBRHSA-N
(2R,5S,6S)-3,3-dimethyl-6-[[(2R)-2-[(3-methylsulfonyl-2-oxoimidazolidine-1-carbonyl)amino]-2-phenylacetyl]amino]-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;sodium
C21H25N5NaO8S2
MFCD07793332
123133909
CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)NC(=O)N4CCN(C4=O)S(=O)(=O)C)C(=O)O)C.[Na]
Specifications
59798-30-0
100mg
MFCD07793332
SGVORSZSYKDVFT-KZHFGBRHSA-N
(2R,5S,6S)-3,3-dimethyl-6-[[(2R)-2-[(3-methylsulfonyl-2-oxoimidazolidine-1-carbonyl)amino]-2-phenylacetyl]amino]-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;sodium
123133909
Mezlocillin sodium salt
White to Yellow
C21H25N5NaO8S2
Soluble in water or DMSO
CC1(C(N2C(S1)C(C2=O)NC(=O)C(C3=CC=CC=C3)NC(=O)N4CCN(C4=O)S(=O)(=O)C)C(=O)O)C.[Na]
562.568
561.56
Product Suggestions
Product Suggestions
Videos
SDS Documents