CAS RN 578-86-9
CAS RN 578-86-9
IUPAC Name:
(2S)-7-hydroxy-2-(4-hydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-4-one
Synonyms:
5-deoxyflavanone (2S)-liquiritigenin liquiritigenin 4',7-dihydroxyflavanone
Molecular Weight (g/mol):
256.26
Molecular Formula:
C15H12O4
InChi Key:
FURUXTVZLHCCNA-AWEZNQCLSA-N
SMILES:
OC1=CC=C(C=C1)[C@@H]1CC(=O)C2=CC=C(O)C=C2O1
1
–
2
of
2
results
2
Liquiritigenin, MedChemExpress
MedChemExpress Liquiritigenin, a flavanone isolated from Glycyrrhiza uralensis, is a highly selective estrogen receptor β (ERβ) agonist with an EC50 of 36.5 nM for activation of the ERE tk-Luc.