CAS RN 57717-80-3
CAS RN 57717-80-3
IUPAC Name:
2,6-di-tert-butyl-4-(3-hydroxy-2,2-dimethylpropyl)phenol
Synonyms:
2,6-di-tert-butyl-4-(3-hydroxy-2,2-dimethylpropyl)phenol
Molecular Weight (g/mol):
292.46
Molecular Formula:
C19H32O2
InChi Key:
XLWJPQQFJNGUPA-UHFFFAOYSA-N
SMILES:
CC(C)(CO)CC1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C