accessibility menu, dialog, popup

UserName

CAS RN 568565-86-6

NH2OFHCl

CAS RN 568565-86-6

IUPAC Name: 1-[4-(4-fluorophenoxy)phenyl]methanamine hydrochloride
Synonyms: 1-[4-(4-fluorophenoxy)phenyl]methanamine hydrochloride
Molecular Weight (g/mol): 253.7
Molecular Formula: C13H13ClFNO
InChi Key: FSCHAOUFEVMQOV-UHFFFAOYSA-N
SMILES: Cl.NCC1=CC=C(OC2=CC=C(F)C=C2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results