CAS RN 568-63-8
CAS RN 568-63-8
IUPAC Name:
disodium 2-(2,4,5,7-tetraiodo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
Synonyms:
erythrosine
Molecular Weight (g/mol):
879.86
Molecular Formula:
C20H6I4Na2O5
InChi Key:
IINNWAYUJNWZRM-UHFFFAOYSA-L
SMILES:
[Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=C(I)C(=O)C(I)=C2OC2=C(I)C([O-])=C(I)C=C12
1
–
2
of
2
results
1
Erythrosin B, ∽93% by uv-vis, MP Biomedicals™
CAS: 568-63-8 Molecular Formula: C40H14I8Na2O10 Molecular Weight (g/mol): 1715.757 MDL Number: MFCD00144257 InChI Key: DKRYETBOLGDIOK-UHFFFAOYSA-L Synonym: Acid Red 51,C.I. 45430 PubChem CID: 131676154 IUPAC Name: disodium;2-(3-hydroxy-2,4,5,7-tetraiodo-6-oxoxanthen-9-yl)benzoic acid;2-(2,4,5,7-tetraiodo-3-oxido-6-oxoxanthen-9-yl)benzoate SMILES: C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)I)O)I)I)I)C(=O)O.C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)I)[O-])I)I)I)C(=O)[O-].[Na+].[Na+]