missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Erythrosin B Sodium Salt, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| CAS | 568-63-8 |
| Molecular Formula | C40H14I8Na2O10 |
| Synonym | Acid Red 51, Solvent Red 140, 2',4',5',7'-Tetraiodofluorescein, FD&C Red No. 3, Erythrosin extra bluish |
| Solubility Information | Soluble in water (20mg/mL) or ethanol (20mg/mL) |
| InChI Key | DKRYETBOLGDIOK-UHFFFAOYSA-L |
| SMILES | C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)I)O)I)I)I)C(=O)O.C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)I)[O-])I)I)I)C(=O)[O-].[Na+].[Na+] |
| IUPAC Name | disodium;2-(3-hydroxy-2,4,5,7-tetraiodo-6-oxoxanthen-9-yl)benzoic acid;2-(2,4,5,7-tetraiodo-3-oxido-6-oxoxanthen-9-yl)benzoate |
| Molecular Weight (g/mol) | 1715.757 |
| PubChem CID | 131676154 |
| Formula Weight | 879.9 |
| Show More |
Safety and Handling
Product Identifier
- Erythrosin B Sodium Salt
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 4
- H302-Harmful if swallowed.
- H413-May cause long lasting harmful effects to aquatic life.
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction