CAS RN 565441-56-7
CAS RN 565441-56-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Bis[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(0), Thermo Scientific Chemicals
CAS: 565441-56-7 Molecular Formula: C96H94Fe2P2Pd Molecular Weight (g/mol): 1527.87 MDL Number: MFCD15071401 Synonym: Pd(Qhos)2 IUPAC Name: Bis[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(0) SMILES: [Fe].[Fe].[Pd].CC(C)(C)P(c1cccc1)C(C)(C)C.CC(C)(C)P(c1cccc1)C(C)(C)C.C1=CC=C(C=C1)c1c(c(c(c1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)c1c(c(c(c1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1