missing translation for 'onlineSavingsMsg'
Learn More

Bis[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(0), Thermo Scientific Chemicals

Product Code. 15465005
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15465005 250 mg 250mg
15427125 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15465005 Supplier Thermo Scientific Alfa Aesar Supplier No. 045913.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Bis[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(0) is used to prepare palladium(0) catalysts for highly selective cross-coupling reactions especially carbohalogenation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 565441-56-7
Molecular Formula C96H94Fe2P2Pd
Molecular Weight (g/mol) 1527.87
MDL Number MFCD15071401
Synonym Pd(Qhos)2
IUPAC Name Bis[1,2,3,4,5-pentaphenyl-1'-(di-tert-butylphosphino)ferrocene]palladium(0)
SMILES [Fe].[Fe].[Pd].CC(C)(C)P(c1cccc1)C(C)(C)C.CC(C)(C)P(c1cccc1)C(C)(C)C.C1=CC=C(C=C1)c1c(c(c(c1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)c1c(c(c(c1C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Pink to White
Quantity 250 mg
Sensitivity Air sensitive
Solubility Information Insoluble in water.
Formula Weight 1527.86
Physical Form Crystalline
Chemical Name or Material Bis[1,2,3,4,5-pentaphenyl-1'- (di-tert-butylphosphino)ferrocene]palladium (0)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.