accessibility menu, dialog, popup

UserName

CAS RN 56441-55-5

H3COOO

CAS RN 56441-55-5

IUPAC Name: ethyl 4-(benzyloxy)benzoate
Synonyms: ethyl 4-(benzyloxy)benzoate
Molecular Weight (g/mol): 256.3
Molecular Formula: C16H16O3
InChi Key: UMOKJIWMQFEFJE-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=C(OCC2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results