missing translation for 'onlineSavingsMsg'
Learn More

Netilmicin sulfate, MedChemExpress

Product Code. 16484177
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16484177 50 mg 50mg
16460728 5 mg 5mg
16458026 100 mg 100mg
16408298 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16484177 Supplier Medchem Express Supplier No. HYA0086
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Netilmicin (sulfate) (SCH-20569 (sulfate)) is an active aminoglycoside antibiotic against most Gram-negative and some Gram-positive bacteria, including certain strains resistant to gentamicin.

Netilmicin (sulfate) (SCH-20569 (sulfate)) is an active aminoglycoside antibiotic against most Gram-negative and some Gram-positive bacteria, including certain strains resistant to gentamicin.

Specifications

Chemical Name or Material Netilmicin sulfate
CAS 56391-57-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H41N5O7.5/2H2SO4
Quantity 50 mg
Synonym SCH-20569 sulfate
Solubility Information H2O : 100 mg/mL (138.74 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (1.39 mM; ultrasonic and warming and heat to 80°C)
SMILES NCC1=CC[C@@H](N)[C@](O[C@@H]2[C@@H](N)C[C@@H](NCC)[C@H](O[C@@]3([H])OC[C@](C)(O)[C@H](NC)[C@H]3O)[C@H]2O)([H])O1.[2.5H2SO4]
Molecular Weight (g/mol) 720.78
Formula Weight 720.78
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.